Learn More
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Shop All Thermo Scientific Products
Click to view available options
Description:
Graphite Liner Sealing Ring
Liner Sealing Ring
Viton Liner Sealing Ring
For Use With (Equipment):
Agilent 2.0 mm Liners
Agilent Direct Straight Liners
Agilent SSL Liners
TRACE 1300 Series GC PTV Injectors
TRACE 1300 Series GC SSL Injectors
TRACE 1300 Series GC and Agilent SSL Injectors
TRACE and FOCUS GC SSL Injectors
Unit Size:
Each
Pack of 10
Pack of 5
Description
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Specifications
Specifications
| Description | Graphite Liner Sealing Ring |
| Quantity | 10 Pack |
| Product Type | Graphite Liner Sealing Ring |
| For Use With (Equipment) | Agilent Direct Straight Liners |