Learn More
Trajan Scientific and Medical™ GC ferrule OGF16-16 one piece graphite 1/16 inch x 1/16 inch fitting size x hole size SGE
Click to view available options
For Use With (Equipment):
1/16 in. Fittings
Unit Size:
Pack of 10
Description
- Easy to use
- Forms reliable seal
- Reusable
- Soft material, forms soft grip to the capillary column
- Low emissions
Specifications
Specifications
| Type | GC Ferrule |
| Description | 1/16 in. I.D. hole |
| Diameter (English) Inside | 1/16 in. |
| Length (Metric) | 3.7 mm |
| Width (Metric) | 3.2 mm |
| For Use With (Equipment) | 1/16 in. Fittings |
| Material | Graphite |
| Quantity | 10/Pk. |
| Diameter (Metric) Inside | 1.59 mm |