Learn More
GBX2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21639-1-AP-150UL
This item is not returnable.
View return policy
Description
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.Specifications
| GBX2 | |
| Polyclonal | |
| Unconjugated | |
| GBX2 | |
| gastrulation brain homeobox 2, GBX2, Homeobox protein GBX 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14472, 2637 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P48031, P52951 | |
| Gbx2 | |
| GBX2 Fusion Protein Ag16288 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |