Learn More
GB/T 24800.2-2009 41 Glucocorticoids 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| CAS | 2135-17-3, 66734-13-2, 51022-69-6, 4419-39-0, 9-8-5534, 378-44-9 |
| Quantity | 1.5 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |