Learn More
BRAND™ Gay-Lussac Uncalibrated Density Bottles
Product Code. 11888271 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11888271 5 mL Pack of 2
11818291 25 mL Pack of 2
11838291 100 mL Pack of 2
11828291 50 mL Pack of 2
11808291 10 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11888271 Supplier BRAND™ Supplier No. 43205
  • ONLINE EXCLUSIVE PRICE
    745.00 NOK
    685.35 NOK / Pack of 2
    Save 59.65 NOK
    8% Off
Estimated Shipment: 10-12-2026
Gay-Lussac density bottle made with Borosilicate Glass 3.3 that is ideal in most research, scientific, and other laboratory environments.
  • BRAND™ density bottle
  • Gay-Lussac pattern
  • Uncalibrated capacity, 10 mL
  • Borosilicate glass, bottle narrow-mouth, bottle round (Gay-Lussac), white bottle
  • DIN ISO 3507
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2Each
Capacity (Metric) 5 mL
Material Borosilicate Glass (3.3)
Product Type Gay-Lussac Density Bottle