Learn More
GATA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11103-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16897953
  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
Description
GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
Specifications
Specifications
GATA2
Polyclonal
Unconjugated
GATA2
GATA binding protein 2, GATA2, NFE1B
Rabbit
Antigen Affinity Chromatography
RUO
14461, 25159, 2624
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O09100, P23769, Q924Y4
Gata2
GATA2 Fusion Protein Ag1557
150 μL
Primary
Human, Mouse, Rat