Learn More
GAT1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28488-1-AP-20UL
 View more versions of this product
Product Code. 16846165
  • 1710.33 NOK / 20µL
Description
The GAT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GAT1. This antibody recognizes human, mouse, rat antigen. The GAT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GAT1
Polyclonal
Unconjugated
20 μL
6529
IgG
Western Blot, Immunohistochemistry, ELISA
467 μg/mL
Rabbit
Primary
Human, Rat, Mouse