Learn More
Thermo Scientific™ Manual, Fixed Needle, Gas Tight Syringes for GC Instruments
Product Code. 13416876 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
Needle Gauge:
22 G
22s G
23 G
23s G
25 G
26 G
26s G
Length (Metric) Needle:
50 mm
57 mm
85 mm
Needle Point Style:
Bevel
Blunt
Cone
Side Hole
Unit Size:
Each
Product Code. Volume (Metric) Needle Gauge Length (Metric) Needle Needle Point Style unitSize
13416876 250 μL 26 G 57 mm Cone Each
12826036 25 μL 23s G 57 mm Cone Each
13486856 25 μL 26s G 85 mm Cone Each
12836036 25 μL 26s G 57 mm Cone Each
12846036 50 μL 23s G 57 mm Cone Each
13436866 50 μL 26s G 85 mm Cone Each
13426876 250 μL 26 G 85 mm Cone Each
10177754 50 μL 25 G 50 mm Bevel Each
13446876 250 μL 23 G 85 mm Side Hole Each
10724457 100 μL 25 G 50 mm Bevel Each
10239214 250 μL 25 G 50 mm Bevel Each
10782317 500 μL 25 G 50 mm Bevel Each
10025774 1 mL 22 G 50 mm Bevel Each
15254573 10 μL 26s G 50 mm Blunt Each
15284573 25 μL 22s G 50 mm Blunt Each
15294573 50 μL 22s G 50 mm Blunt Each
15204583 100 μL 22s G 50 mm Blunt Each
15214583 500 μL 22s G 50 mm Blunt Each
18 options
Product Code. 13416876
Brand: Thermo Scientific™ 365I2331
Choose Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for manual accurate, reproducible GC sample introduction using an affixed needle and a plunger that creates a tight seal.
Get durability, clarity and accuracy from Thermo Scientific™ Manual Fixed Needle, Gas Tight Syringes. Used for manual sample injection into gas chromatography (GC) instrument, they provide accuracy and reproducibility that gives you confidence in analytical results, time-after-time. These syringes have an affixed needle and a plunger that creates a tight seal with the barrel. Choose from a variety of sizes and types. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
Durability
The enhanced design of the syringes delivers:
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
Clarity
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
Accuracy
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover:
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
TRUSTED_SUSTAINABILITY
Specifications
Description 250 μL; 57 mm L; 26 gauge fixed
Needle Gauge 26 G
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Quantity Ea.
Volume (Metric) 250 μL
Length (Metric) Needle 57 mm
Needle Point Style Cone
For Use With (Equipment) TriPlus RSH Autosampler