Learn More
Cole-Parmer™ Gast Oilles Air Compressor
Product Code. 15794598 Shop All Cole-Parmer Products
Change view
Click to view available options
Motor Speed:
0 Hp
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Motor Speed unitSize
15794598 0 Hp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15794598 Supplier Cole-Parmer™ Supplier No. 0705415
This item requires a price update prior to being ordered.
Request a Price for this item here.
Diaphragm air compressors offer oil-free, cool air, even at maximum duty. Air entering the pump does not go near the motor to pick up heat.
  • Diaphragm compressor pump
  • Virtually maintenance free
  • Ideal for light duty applications
Specifications
Amperage 0.4 amp
Includes Power cord without plug
Motor Speed 0 Hp
Description Oilless air compressor, diaphragm compressor pump 1.1 cfm
Material PTFE, Stainless Steel, and Aluminum Wetted Parts
Model DOA-P501-BN
Temperature (English) Max. 100°F
Temperature (Metric) Max 38°C
Overall Height (English) 7.625 in.
Overall Height (Metric) 19.37 cm
Overall Length (English) 7.625 in.
Overall Length (Metric) 19.37 cm
Overall Width (English) 5.125 in.
Overall Width (Metric) 13.2 cm
Maximum Pressure 60 psi
Voltage 220/240 V