Learn More
BD GasPak™ Supplies
Product Code. 11713473
Change view
Click to view available options
Type:
CO2 Indicator Strip
Catalyst Replacement Charges
Dry Anaerobic Indicator Strip
Tube Holder
Quantity:
1 Each
10 Pk.
100 Pk.
50 Pk.
Unit Size:
Each
Pack of 100
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity unitSize
11713473 CO2 Indicator Strip 50 Pk. Pack of 50
11703473 Dry Anaerobic Indicator Strip 100 Pk. Pack of 100
15930997 Catalyst Replacement Charges 10 Pk. Each
10252705 Tube Holder 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11713473 Supplier BD Supplier No. 271055
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.
GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Accessories include:
  • CO2 Indicator Strips
  • Dry Anaerobic Indicator Strips
  • Tube Holder
  • Catalyst Replacement Charges
  • Catalyst Reaction Chamber
TRUSTED_SUSTAINABILITY
Specifications
Type CO2 Indicator Strip
Content And Storage 15°C to 30°C
Description Indicator signifies that the carbon dioxide concentration in the atmosphere is 3% or greater.
Format Strip
For Use With (Application) Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria
For Use With (Equipment) BD GasPak™
Quantity 50 Pk.