Learn More
BD GasPak™ 150 Replacement Parts and Accessories
Product Code. 11729022
Change view
Click to view available options
Type:
Anaerobic Jar
Inner Lid
Lid Assembly
Petri Dish Rack
Unit Size:
Each
Product Code. Type unitSize
11729022 Lid Assembly Each
15991652 Inner Lid Each
12340439 Anaerobic Jar Each
11739022 Petri Dish Rack Each
4 options
Product Code. 11729022
Brand: BD 260610
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Lid Assembly
For Use With (Equipment) GasPak™ 150
Includes Outer lid and thumbscrew, inner lid, large O-ring gasket, 3 catalyst reaction chambers, 6 charges