missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GASP-1/WFIKKN2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF2070-SP
This item is not returnable.
View return policy
Description
GASP-1/WFIKKN2 Polyclonal specifically detects GASP-1/WFIKKN2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
| GASP-1/WFIKKN2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GASP1, GASP-1, Growth and differentiation factor-associated serum protein 1, hGASP-1, WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-relatedprotein, WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2, WAP, FS, Ig, two KU and NTR module related protein, WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2, WFIKKN2, WFIKKN-related protein, WFIKKNRP, WFIKKNRPmultivalent protease inhibitor protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 124857 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 18.9 ug/mL | |
| Q8TEU8 | |
| WFIKKN2 | |
| Mouse myeloma cell line NS0-derived recombinant human GASP-1/WFIKKN2 Leu35-His576 Accession # Q8TEU8 | |
| 25 μg | |
| Primary | |
| Detects human GASP-1/WFIKKN2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human GASP-2 is observed. | |
| Human | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?