missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13795-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| GAS6 | |
| Polyclonal | |
| Unconjugated | |
| GAS6 | |
| AXLLG, AXSF, GAS 6, GAS6, growth arrest 6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2621 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14393 | |
| GAS6 | |
| GAS6 Fusion Protein Ag4617 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction