Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottle Head without Filter Disc
Description
- Acc. to DIN 12596, with cone
- For bottles acc. to DIN 12463
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Material | Duran Tubing |
| Type | Gas Washing Bottle Head without Filter Disc |
| Includes | Glass Olives |
| Standard Taper Joint | NS 29/32 |
| Diameter (Metric) Outside | 11 mm |
| Certifications/Compliance | DIN 12596 |
| Quantity | 1 Pack |