Learn More
Lenz Laborglasintrumente™ Gas Washing Bottle Head without Filter Disc
Gas washing head for bottles acc. to DIN 12463
Brand: Lenz Laborglasintrumente™ 05500610
Additional Details : Weight : 0.10000kg
Description
- Acc. to DIN 12596
- With cone NS 29/32
- Material: DURAN™ tubing
Specifications
With thread connections (GL 14): 11 mm | |
Gas washing bottles acc. to DIN 12463 | |
1 |