Description
- Distribute to up to 5 positions or vessels.
- Does not control or regulate gas/vacuum flow.
- Leak-proof shut-off valves.
Specifications
Specifications
| For Use With (Equipment) | Starfish Magnetic Stirrer System. |
| For Use With (Application) | Gas/Vacuum Distribution. |
| Includes | Leak-proof shut-off valves. |