Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Cone Tip
Product Code. p-8397064
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
500 μL
500 mL
Diameter (Metric) Inner Needle:
0.11 mm
0.20 mm
0.32 mm
Diameter (Metric) Outer Needle:
0.47 mm
0.63 mm
Length (Metric) Needle:
57 mm
85 mm
Unit Size:
Each
Product Code. Volume (Metric) Diameter (Metric) Inner Needle Diameter (Metric) Outer Needle Length (Metric) Needle unitSize
13126105 500 mL 0.32 mm 0.63 mm 57 mm Each
13136105 500 μL 0.32 mm 0.63 mm 85 mm Each
13139215 100 μL 0.11 mm 0.63 mm 57 mm Each
13146105 500 μL 0.20 mm 0.47 mm 57 mm Each
13149215 100 μL 0.11 mm 0.63 mm 85 mm Each
13156105 500 μL 0.20 mm 0.47 mm 85 mm Each
13159215 100 μL 0.11 mm 0.47 mm 57 mm Each
13166095 250 μL 0.32 mm 0.63 mm 57 mm Each
13169215 100 μL 0.11 mm 0.47 mm 85 mm Each
13176095 250 μL 0.32 mm 0.63 mm 85 mm Each
13186095 250 μL 0.20 mm 0.47 mm 57 mm Each
13196095 250 μL 0.20 mm 0.47 mm 85 mm Each
12 options
Product Code. 13126105
Brand: Trajan Scientific and Medical™ 007862
Designed to provide improved solvent resistance, greater temperature range and increased operational smoothness.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Material PTFE-tipped (Plunger)
Needle Gauge 23 G
Diameter (Metric) Inner Needle 0.32 mm
Diameter (Metric) Outer Needle 0.63 mm
Product Type Gastight
Volume (Metric) 500 mL
Length (Metric) Needle 57 mm
Needle Point Style Cone
Connection Type Fixed