Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Cone Tip
Product Code. 13126105
Click to view available options
Volume (Metric):
100 μL
250 μL
500 μL
500 mL
Diameter (Metric) Inner Needle:
0.11 mm
0.20 mm
0.32 mm
Diameter (Metric) Outer Needle:
0.47 mm
0.63 mm
Length (Metric) Needle:
57 mm
85 mm
Unit Size:
Each
Product Code. 13126105
Brand: Trajan Scientific and Medical™ 007862
Designed to provide improved solvent resistance, greater temperature range and increased operational smoothness.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Material PTFE-tipped (Plunger)
Needle Gauge 23 G
Diameter (Metric) Inner Needle 0.32 mm
Diameter (Metric) Outer Needle 0.63 mm
Product Type Gastight
Volume (Metric) 500 mL
Length (Metric) Needle 57 mm
Needle Point Style Cone
Connection Type Fixed