Learn More
Lenz Laborglasintrumente Gas Inlet Tubes, Straight
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05080002
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11836114
  • 190.80 NOK / Each
Estimated Shipment: 26-06-2024
Description
  • Straight air leak tubes
  • Built-in length: 200 mm
  • With NS cone and thin capillary (easily to draw by burner)
  • Made of DURAN™ tubing
Specifications
Specifications
NS 19/26