Learn More
OHAUS™ Gas Cylinder Support with Chain
Product Code. 15712449 Shop All Ohaus Products
Click to view available options
Includes:
Message Strap
Strap
Type:
Bench Support
Wall Support
Unit Size:
Each
Product Code. 15712449
Brand: OHAUS™ 30400029
Only null left
Add to basket
Add to basket
OHAUS™ Gas Cylinder Support with Chain provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Type Wall Support
Material Cast Aluminum
Includes Message Strap
For Use With (Application) Gas Cylinder Support
Description Clamp, Support, 717M Chain, CLR-717M
Height (Metric) 118 mm
Height (English) 4.63 in.
Length (English) 1.88 in.
Length (Metric) 48 mm
Width (English) 8.10 in.
Width (Metric) 206 mm