Learn More
Thermo Scientific™ Gas Chromatography Tubing
Product Code. 15294603 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
Copper
Stainless Steel
Diameter (English) Inner:
0.01 in.
0.02 in.
0.03 in.
0.04 in.
0.065 in.
0.085 in.
0.19 in.
0.21 in.
Diameter (English) Outer:
1/16 in.
1/4 in.
1/8 in.
Description:
25 ft. Length
50 ft. Length
Unit Size:
Each
Product Code. Material Diameter (English) Inner Diameter (English) Outer Description unitSize
15294603 Stainless Steel 0.04 in. 1/16 in. 25 ft. Length Each
15244603 Copper 0.065 in. 1/8 in. 50 ft. Length Each
15254603 Copper 0.19 in. 1/4 in. 50 ft. Length Each
15264603 Stainless Steel 0.01 in. 1/16 in. 25 ft. Length Each
15274603 Stainless Steel 0.02 in. 1/16 in. 25 ft. Length Each
15284603 Stainless Steel 0.03 in. 1/16 in. 25 ft. Length Each
15204613 Stainless Steel 0.085 in. 1/8 in. 25 ft. Length Each
15214613 Stainless Steel 0.21 in. 1/4 in. 25 ft. Length Each
8 options
Product Code. 15294603
Brand: Thermo Scientific™ 60181637
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Find the copper and stainless steel tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Find copper and stainless steel Thermo Scientific™ Gas Chromatography Tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Gas Chromatography Tubing
Material Stainless Steel
Description 25 ft. Length
Diameter (English) Inner 0.04 in.
Diameter (English) Outer 1/16 in.
For Use With (Equipment) Gas Chromatography Systems