Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Device
Product Code. 15297508 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1/Ea.
Unit Size:
Each
Product Code. Quantity unitSize
15297508 1/Ea. Each
1 options
Product Code. 15297508
Brand: Thermo Scientific™ 60201398
Only null left
Add to basket
Add to basket
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device.
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device. This device comprises a gas module with microfluidics and Thermo Scientific™ SilTite™ FingerTite fittings for easy set up and a reliable seal.
TRUSTED_SUSTAINABILITY
Specifications
Type 3-Port SilFlow MCD
Description 3-Port SilFlow MCD (0.25/0.32)
Quantity 1/Ea.
For Use With (Equipment) Gas Chromatography Systems