missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15831-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. The encoded enzyme is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.Specifications
| GARS | |
| Polyclonal | |
| Unconjugated | |
| GARS | |
| AP 4 A synthetase, CMT2D, DSMAV, GARS, Glycine tRNA ligase, glycyl tRNA synthetase, GlyRS, HMN5, SMAD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2617, 297113, 353172 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P41250, Q5I0G4, Q9CZD3 | |
| Gars | |
| GARS Fusion Protein Ag8322 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction