missing translation for 'onlineSavingsMsg'
Learn More

GARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16855194 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16855194 20 μL 20µL
16845194 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16855194

Brand: Proteintech 158311AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. The encoded enzyme is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.

Specifications

Antigen GARS
Applications Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GARS
Gene Accession No. P41250, Q5I0G4, Q9CZD3
Gene Alias AP 4 A synthetase, CMT2D, DSMAV, GARS, Glycine tRNA ligase, glycyl tRNA synthetase, GlyRS, HMN5, SMAD1
Gene Symbols Gars
Host Species Rabbit
Immunogen GARS Fusion Protein Ag8322
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2617, 297113, 353172
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.