Learn More
GAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16852884 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16852884 20 μL 20µL
16842884 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16852884 Supplier Proteintech Supplier No. 143051AP20UL
Rabbit Polyclonal Antibody
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
Specifications
Antigen GAN
Applications Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GAN
Gene Accession No. Q8CA72, Q9H2C0
Gene Alias GAN, GAN1, gigaxonin, Kelch like protein 16, KLHL16
Gene Symbols GAN
Host Species Rabbit
Immunogen GAN Fusion Protein Ag5421
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209239, 307893, 8139
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG