missing translation for 'onlineSavingsMsg'
Learn More

GAMT Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E7B11, Proteintech

Product Code. 16898085 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16898085 20 μL 20µL
16888085 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16898085

Brand: Proteintech 663221IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GAMT is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in its gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals.The protein encoded by this gene is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in this gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals. Two transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen GAMT
Applications Western Blot
Classification Monoclonal
Clone 1E7B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GAMT
Gene Accession No. O35969, P10868, Q14353
Gene Alias GAMT, PIG2, TP53I2
Gene Symbols GAMT
Host Species Mouse
Immunogen GAMT Fusion Protein Ag16778
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100625146, 14431, 25257, 2593
Target Species Pig, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.