missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Synuclein Antibody (SAIC-31A-10) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20099JF585
This item is not returnable.
View return policy
Description
gamma-Synuclein Monoclonal antibody specifically detects gamma-Synuclein in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Mass Spectrometry,Immunocytochemistry/ Immunofluorescence
Specifications
| gamma-Synuclein | |
| Monoclonal | |
| Janelia Fluor 585 | |
| BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1) | |
| The KLH-conjugated peptide 'YLAPSGPSGTLK', derived from Gamma-synuclein. Gamma-synulein plays a role in neurofilament network integrity. | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cellular Markers, Core ESC Like Genes, Mitotic Regulators, Neuroscience, Stem Cell Markers | |
| 6623 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, ELISA, Western Blot, Mass Spectrometry, Immunocytochemistry/Immunofluorescence | |
| SAIC-31A-10 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction