Learn More
Thermo Scientific™ gamma-Secretase Inhibitor II
Product Code. 15486599
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 15486599
Brand: Thermo Scientific™ J64828.M^
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A cell-permeable, reversible and selective peptidomimetic inhibitor of --secretase
Molecular Formula C20H30N2O4
Formula Weight 362.47
Synonym MW167
Product Type gamma-Secretase Inhibitor
Form Solid