Learn More
Keystone Cleanroom Products Gamma-Irradiated Heat-Seal Sterility Maintenance Bag
GREENER_CHOICE
Product Code. 17706889 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
25.4 x 15.2 cm (6 x 10 in.)
30.5 x 20.3 cm (8 x 12 in.)
38.1 x 25.4 cm (10 x 15 in.)
50.8 x 30.5 cm (12 x 20 in.)
56 x 40.6 cm (16 x 22 in.)
76.2 x 40.6 cm (16 x 30 in.)
76.2 x 61 cm (24 x 30 in.)
94 x 63.5 cm (25 x 37 in.)
Unit Size:
Pack of 250
Pack of 500
Product Code. Dimensions (L x W) unitSize
17706889 94 x 63.5 cm (25 x 37 in.) Pack of 250
17736879 25.4 x 15.2 cm (6 x 10 in.) Pack of 500
17746879 30.5 x 20.3 cm (8 x 12 in.) Pack of 500
17756879 38.1 x 25.4 cm (10 x 15 in.) Pack of 500
17766879 50.8 x 30.5 cm (12 x 20 in.) Pack of 500
17776879 56 x 40.6 cm (16 x 22 in.) Pack of 250
17786879 76.2 x 40.6 cm (16 x 30 in.) Pack of 250
17796879 76.2 x 61 cm (24 x 30 in.) Pack of 250
8 options
Product Code. 17706889
Brand: Keystone Cleanroom Products 3235ST
Bag used to cover wrapped items after sterilization to provide protection from environmental factors that could compromise sterility.
  • Made of a transparent, strong, 3 mil thick medical grade polyolefin material
  • Features a starter slit to facilitate easy opening of the cover without the need for cutting instruments which could damage the contents
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 27.4 L
Certifications/Compliance COC, Certificate of Irradiation
Closure Type Heat Seal
Color Clear
Disposable Single-use
Material Polyolefin
Sterility Sterile
Thickness (Metric) 76.2 μm
Latex Free Yes
Thickness (English) 3 mil
Dimensions (L x W) 94 x 63.5 cm (25 x 37 in.)
Length (English) 37 in.
Length (Metric) 94 cm
Width (English) 25 in.
Width (Metric) 63.5 cm
Product Type Cover