Learn More
Usbeck Laborgeraete™ Galvanized Steel Tripod
Product Code. p-8010261
Click to view available options
Height (Metric):
180 mm
210 mm
230 mm
Unit Size:
Each
Product Code. 10260931
Brand: Usbeck Laborgeraete™ 5132
Only null left
Specifications
Material Galvanized Steel
Height (Metric) 210 mm
Diameter (Metric) Inner 120 mm
Product Type Tripod
Quantity 1 Ea.
Weight (Metric) 340 g