Learn More
Usbeck Laborgeraete™ Galvanized Steel Tripod
Product Code. 10137011
Change view
Click to view available options
Height (Metric):
180 mm
210 mm
230 mm
Unit Size:
Each
Product Code. Height (Metric) unitSize
10137011 230 mm Each
10260931 210 mm Each
10600293 180 mm Each
3 options
Product Code. 10137011
Brand: Usbeck Laborgeraete™ 5134
Only null left
Specifications
Material Galvanized Steel
Height (Metric) 230 mm
Diameter (Metric) Inner 160 mm
Product Type Tripod
Quantity 1 Ea.
Weight (Metric) 400 g