Learn More
GALK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15284-1-AP-150UL
This item is not returnable.
View return policy
Description
Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts in the adult population. GALK1 sequence shares the greatest level of conservation, 44.5% identity with that from E. coli and 34.6% amino acid identity with the product of the human GALK2 gene.Specifications
| GALK1 | |
| Polyclonal | |
| Unconjugated | |
| GALK1 | |
| Galactokinase, galactokinase 1, Galactose kinase, GALK, GALK1, GK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14635, 2584, 287835 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51570, Q9R0N0 | |
| Galk1 | |
| GALK1 Fusion Protein Ag7403 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |