missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-10 Antibody (561603) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5447JF525
This item is not returnable.
View return policy
Description
Galectin-10 Monoclonal antibody specifically detects Galectin-10 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
Specifications
| Galectin-10 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Charcot-Leyden crystal proteinLGALS10A, EC 3.1.1.5, eosinophil lysophospholipase, Gal-10, GAL10, galectin 10, galectin-10, LGALS10, LPPL_HUMAN, Lysolecithin acylhydrolase, MGC149659 | |
| E. coli-derived recombinant human Galectin-10, Met1-Arg142, Accession # Q05315 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
| 561603 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Signal Transduction | |
| 1178 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction