Learn More
Eppendorf™ Galaxy™ 48 R
Description
- Provides mid-sized capacity within a small footprint
- Fanless design offers more usable space
- Six-sided direct heating system provides a uniform incubated environment
- Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
- Low gas consumption
- Integrated alarm relay
Specifications
Specifications
| Capacity (Metric) | 48 L |
| Oxygen Control | 1 to 19 % |
| Temperature Range (Metric) | Ambient +4° to 50°C |
| Display | Standard display |
| Includes | No High Temp Disinfection |
| Ports | 1 |
| Dimensions (D x W x H) Interior | 30.5 x 401 x 40.1 cm |