Learn More
GAL3ST4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16240-1-AP-20UL
 View more versions of this product
Product Code. 16895804
  • 1710.33 NOK / 20µL
Description
Catalyzes the transfer of sulfate to beta-1,3-linked galactose residues in O-linked glycoproteins. Good substrates include asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc.
Specifications
Specifications
GAL3ST4
Polyclonal
Unconjugated
GAL3ST4
GAL3ST 4, GAL3ST4
Rabbit
Antigen Affinity Chromatography
RUO
330217, 498166, 79690
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96RP7
Gal3st4
GAL3ST4 Fusion Protein Ag9063
20 μL
Primary
Mouse, Human, Rat