missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD2 / GAD65 (GABAergic Neuronal Marker) Mouse, Clone: GAD2/1960, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2572-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
GAD2 / GAD65 (GABAergic Neuronal Marker) | |
Cytoplasmic | |
GAD2/1960 | |
Unconjugated | |
Mouse | |
65kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
2572 | |
IgG2b κ |
ELISA, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human GAD2 (GAD65) protein fragment (apprx. aa 6-99) | |
Pancreas or Brain. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |