Learn More
Gabapentin, TRC
Product Code. 30390649
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390649 50 mg -
30389213 10 mg -
30399030 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30390649 Supplier Toronto Research Chemicals Supplier No. TRCG11725050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60142-96-3
Molecular Formula C9 H17 N O2
Molecular Weight (g/mol) 171.24
Synonym 1-(Aminomethyl)-cyclohexaneacetic acid, CI 945, GOE 2450, GOE 3450, Gabapen, Gabapentin, Go 3450, Neurontin
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
SMILES NCC1(CC(=O)O)CCCCC1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)
Formula Weight 171.1259
Percent Purity >95%
Chemical Name or Material Gabapentin
Grade HPLC