Learn More
Gabapentin enacarbil, MedChemExpress
Product Code. 16499547
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16499547
Brand: Medchem Express HY16216
MedChemExpress Gabapentin enacarbil (XP-13512) is a prodrug for the anticonvulsant and analgesic drug gabapentin.
Gabapentin enacarbil (XP-13512) is a prodrug for the anticonvulsant and analgesic drug gabapentin.
Specifications
Chemical Name or Material Gabapentin enacarbil
Physical Form Viscous Liquid
CAS 478296-72-9
Color Light Brown
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H27NO6
For Use With (Application) Neuroscience-Neuromodulation
Synonym XP-13512
Solubility Information Ethanol : 100 mg/mL (303.59 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (303.59 mM) ∣H2O : 0.67 mg/mL (2.03 mM; Need ultrasonic)
SMILES O=C(O)CC1(CNC(OC(OC(C(C)C)=O)C)=O)CCCCC1
Molecular Weight (g/mol) 329.39
Formula Weight 329.39
Percent Purity 95.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.