Learn More
Gabapentin (1.0 mg/mL in Methanol), TRC
Product Code. 30404588
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30404588 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30404588 Supplier Toronto Research Chemicals Supplier No. TRCKIT123510X1ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 60142-96-3
Molecular Formula C9 H17 N O2
Quantity 10 x 1 mL
Synonym 1-(Aminomethyl)-cyclohexaneacetic acid, CI 945, GOE 2450, GOE 3450, Gabapen, Gabapentin, Go 3450, Neurontin
InChI Formula InChI=1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)
SMILES NCC1(CC(=O)O)CCCCC1
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
Molecular Weight (g/mol) 171.24
Formula Weight 171.1259
Concentration or Composition (by Analyte or Components) Gabapentin 1 mg/mL
Chemical Name or Material Gabapentin