missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 4 Antibody (S398A-34), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59326B
This item is not returnable.
View return policy
Description
GABA-AR alpha 4 Monoclonal specifically detects GABA-AR alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GABA-AR alpha 4 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GABRA4 | |
| Fusion protein amino acids 410-450 (Cytoplasmic C-terminus) of rat GABA-A-Receptor Alpha 4 | |
| 100 μL | |
| Primary | |
| Detects 60kDa. Does not cross-react with other GABA-A-Receptor Alpha subunits. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S398A-34 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| GABA(A) receptor subunit alpha-4, gamma-aminobutyric acid (GABA) A receptor, alpha 4, gamma-aminobutyric acid receptor subunit alpha-4 | |
| Mouse | |
| Protein G purified | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2557 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction