missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-43558
This item is not returnable.
View return policy
Description
Specifically detects GABA in All samples, and it is validated for Dot Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
GABA Polyclonal specifically detects GABA in Human, Mouse, Rat, Drosophila samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.
Specifications
| GABA | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal | |
| GABA | |
| The immunogen used to generate this antibody corresponds to GABA | |
| 0.1 mL | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1mg/mL | |
| Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000, Dot Blot | |
| y-Aminobutyric acid | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction