missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody is specific to GAB3.
GAB3 belongs to the GAB family. GAB3 interacts with PIK3R/p85, SHP2 and GRAP2/MONA. It may interact with Grb2.
Specifications
Specifications
| Antigen | GAB3 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | GAB3 |
| Gene Accession No. | Q8BSM5, Q8WWW8 |
| Gene Alias | GAB3, GRB2 associated binder 3 |
| Gene Symbols | Gab3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?