missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20089-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to GAB3.
GAB3 belongs to the GAB family. GAB3 interacts with PIK3R/p85, SHP2 and GRAP2/MONA. It may interact with Grb2.Specifications
| GAB3 | |
| Polyclonal | |
| Unconjugated | |
| GAB3 | |
| GAB3, GRB2 associated binder 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 139716, 210710 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BSM5, Q8WWW8 | |
| Gab3 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction