missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GAB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. p-200128310
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
Product Code. Quantity unitSize
17740574 200 μL 200µL
17703374 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 17740574

Brand: Invitrogen™ PA5119162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GAB3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

GAB3 belongs to the GAB family. GAB3 interacts with PIK3R/p85, SHP2 and GRAP2/MONA. It may interact with Grb2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAB3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GAB3
Gene Accession No. Q8BSM5, Q8WWW8
Gene Alias 5930433H21Rik; DOS/Gab family member 3; GAB3; Gab3 scaffolding protein; GRB2 associated binding protein 3; GRB2-associated binder 3; GRB2-associated binding protein 3; GRB2-associated-binding protein 3; Growth factor receptor bound protein 2-associated protein 3
Gene Symbols GAB3
Host Species Rabbit
Immunogen Recombinant protein GRB2 Associated Binding Protein 3. The antigen corresponds to amino acid range 142-375 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139716, 210710
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.