Learn More
G6PD Mouse anti-Human, Unconjugated, Clone: 2A7B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66373-1-IG-150UL
Description
The G6PD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human G6PD. This antibody recognizes human, mouse, rat antigen. The G6PD antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| G6PD | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2A7B12 | |
| Unconjugated | |
| 150 μL | |
| 2539 | |
| IgG2a |