Learn More
G3BP1 Antibody, Novus Biologicals™
Product Code. 18791324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
Product Code. Quantity unitSize
18791324 0.1 mL 0.1mL
18439800 25 μL 25µL
2 options
Product Code. 18791324
Brand: Novus Biologicals NBP183404
Rabbit Polyclonal Antibody has been used in 1 publication
G3BP1 Polyclonal specifically detects G3BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mL
Regulatory Status RUO
For Research Use Only