Learn More
Asecos™ G-OD Outdoor Safety Storage Cabinet
Product Code. 15516895 Shop All asecos Products
Change view
Click to view available options
Configuration:
1 Rolling Ramp
Tall Cabinet
Door Style:
Wing
Dimensions (D x W x H):
400 x 1006 x 2149 mm
400 x 1356 x 2149 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Door Style Dimensions (D x W x H) unitSize
15516895 Tall Cabinet Wing 400 x 1356 x 2149 mm Each
15709097 1 Rolling Ramp Wing 400 x 1006 x 2149 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15516895 Supplier Asecos™ Supplier No. 30692015
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
For the safe and approved outdoor storage of flammable or toxic gases.
  • Robust, high-quality construction perfect for installation in outdoor areas
  • Completely galvanized sheet steel construction with plastic laminated structured surface and stainless steel base
  • Sloping roof with supernatant allows water to drain
  • Wing doors with a large opening angle for ease of access
  • Solid 3-point locking with lockable turning lever (also available as profile cylinder lock)
  • Effective ventilation through openings beneath the doors and at the rear wall
  • Comfortable adjusting possibilities – integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors
  • In accordance with TRGS 510
Specifications
Certifications/Compliance TRGS 510
Configuration Tall Cabinet
Dimensions (D x W x H) Interior 370 x 1345 x 2070 mm
Dimensions (D x W x H) 400 x 1356 x 2149 mm
Door Style Wing
For Use With (Application) Outdoor storage of gas cylinders
Includes No Window
Load Bearing Capacity 600 kg
No. of Doors 2
Surface Coating Plastic laminated
Weight (Metric) 161 kg
Color Light Gray
Depth (English) Exterior 15.75 in.
Height (English) Exterior 84.61 in.
Material Galvanized Steel
Product Type Safety Storage Cabinet
Width (English) Exterior 53.39 in.