Learn More
FYN Mouse anti-Human, Mouse, Rat, Clone: 1G4B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66606-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16888635
  • 1611.84 NOK / 20µL
Estimated Shipment: 07-05-2024
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.
Specifications
Specifications
FYN
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P06241, P39688, Q62844
Fyn
FYN Fusion Protein Ag11127
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1G4B1
Unconjugated
FYN
FYN, p59 Fyn, Proto oncogene c Fyn, Proto oncogene Syn, SLK, Src like kinase, SYN, Tyrosine protein kinase Fyn
Mouse
Protein G
RUO
14360, 25150, 2534
-20°C