missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
1975.00 NOK - 2985.00 NOK
Specifications
| Antigen | FXR1 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:300-1:10000 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606225
|
Novus Biologicals
NBP2-98797-100ul |
100 μg |
2985.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693446
|
Novus Biologicals
NBP2-98797-50ul |
50 μg |
1975.00 NOK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FXR1 Polyclonal antibody specifically detects FXR1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| FXR1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Immune System Diseases, Immunology | |
| 0.2 um filtered solution in PBS | |
| 8087 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| fragile X mental retardation syndrome-related protein 1, fragile X mental retardation, autosomal homolog 1, FXR1P, hFXR1p | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FXR1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title