Learn More
Invitrogen™ FxCycle™ PI/RNase Staining Solution
Product Code. 13433329
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13433329 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13433329 Supplier Invitrogen™ Supplier No. F10797
  • ONLINE EXCLUSIVE PRICE
    1390.00 NOK
    1042.65 NOK / 100mL
    Save 347.35 NOK
    24% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
FxCycle™ PI/RNase Staining Solution
FxCycle™ PI/RNase Staining Solution is used for flow cyotmetric analysis of DNA content in fixed cells. Propidium iodide (PI), a popular red-fluorescent stain, binds to DNA by intercalating between the bases with little or no sequence preference and with a stoichiometry of one dye per 4–5 base pairs of DNA. PI also binds to RNA, necessitating treatment with RNase to distinguish between RNA and DNA staining. This propidium iodide solution includes DNase-free RNase A and a permeablization reagent. It comes ready to use: simply add the staining solution to fixed cells, incubate, and acquire on a flow cytometer without washing.
Related Links:
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 bottle FxCycle™ PI/RNase Staining Solution (100 mL). Store at room temperature, and protect from light.Store at room temperature
Excitation Wavelength Range 535⁄ 617
Dye Type propidium iodide
Format Tube(s)
For Use With (Equipment) Flow Cytometer
Quantity 100 mL
Detection Method Fluorescence
Form Solution
Product Line FxCycle
Shipping Condition Room Temperature
Product Type Staining Solution
Sub Cellular Localization Nucleus